Learn More
Alkali Scientific Dicloxacillin Sodium Salt, 100 Mg
SDP
Supplier:  Alkali Scientific DIC001CP
Encompass_Preferred
Dicloxacillin Sodium Salt, 100mg, is a semisynthetic penicillin antibiotic used for laboratory research involving antibacterial susceptibility testing. It is effective against a wide range of penicillinase-producing Gram-positive bacteria. Dicloxacillin is commonly employed in experiments to assess the antibacterial properties of various compounds or in combination with other antimicrobial agents. This formulation is used to determine the efficacy of antibiotics in inhibiting bacterial growth in vitro, making it a valuable tool in microbiological research and the development of new antibacterial therapies.
Catalog No. 50-006-60931