Learn More
Strem, An Ascensus Company Dicyclohexylchlorophosphine, min. 98%; [16523-54-9]; (C6H11)2PCl; Mol Wt:232.74; Color & Form: colorless to pale yellow, cloudy liq.; Size: 1g
SDP
Supplier:  Strem, An Ascensus Company 1510501G
Encompass
Dicyclohexylchlorophosphine, min. 98%; [16523-54-9]; (C6H11)2PCl; Mol Wt:232.74; Color & Form: colorless to pale yellow, cloudy liq.; Size: 1g
Catalog No. 50-138-5397