Learn More
Strem, An Ascensus Company CAS# 829-84-5. 10g. Dicyclohexylphosphine, 98%. MFCD00014296
SDP
Supplier:  Strem, An Ascensus Company 15112010G
Encompass
CAS# 829-84-5. 10g. Dicyclohexylphosphine, 98%. MFCD00014296. Molecular Weight: 198.29. Molecular Formula: (C6H11)2PH. Color/form: light orange yellow liq. Strem# 15-1120. http://www.strem.com/catalog/v/15-1120/
Catalog No. 50-901-10076