Learn More
Medchemexpress LLC Dicyclomine impurity 1 | 60263-54-9 | 210.31 g/mol | C13H22O2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW043718500MG
Encompass_Preferred
Dicyclomine impurity 1 is an analytical reference impurity of dicyclomine intended for research and analytical use. It is supplied as a solid standard for identification, method development, and quantitation of related impurities.
  • Intended for research and analytical use.
  • Suitable as a reference standard for impurity profiling and method development.
  • Solid form; store at room temperature as indicated in the datasheet.
  • Available in a 500 MG package.
  • CAS number: 60263-54-9; formula: C13H22O2; molecular weight: 210.31 g/mol.
Catalog No. 50-002-10229