Learn More
Medchemexpress LLC DiD perchlorate | 127274-91-3 | 98.6% | 959.90 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYD10281MG
Encompass_Preferred
DiD perchlorate is a long-chain carbocyanine dye that is broadly utilized to label cells, organelles, liposomes, viruses, and lipoproteins. It is a far-red fluorescent lipophilic cyanine dye that can be used for intravital imaging. The compound is unstable in solutions, so freshly prepared solutions are recommended.
  • Long-chain carbocyanine dye.
  • Used to label cells, organelles, liposomes, viruses, and lipoproteins.
  • Far-red fluorescent lipophilic cyanine dye.
  • Can be used in intravital imaging.
  • Applications include extracellular vesicle (EVs) staining.
  • Used in various research publications, including studies on cellular uptake, tracking of mesenchymal stem cells, and fluorescence imaging in tumor-bearing mice.
Catalog No. 50-004-30663