Learn More
Cayman Chemical DID4chLRobenzensulfonAT 10mg
SDP
Supplier:  Cayman Chemical 2911510 MG
Encompass_Preferred
A lipophilic fluorescent probe; rapidly incorporated into phospholipid cell membranes; has been used to label the plasma membrane and endocytic organelles in bovine aorta endothelial cells and rat hippocampal slices; has been used to assess proliferation in prostate cancer cell lines by flow cytometry; ex/em = 650/670 nm, respectively
Catalog No. 50-293-9693