Learn More
Medchemexpress LLC HY-100662 5mg Medchemexpress, Didesethyl chloroquine CAS:4298-14-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1006625MG
Encompass_Preferred
Medchemexpress, HY-100662 5mg Didesethyl chloroquine CAS:4298-14-0 Didesethyl chloroquine (Bisdesethylchloroquine) is a major metabolite of the antimalarial drug Chloroquine. Didesethyl chloroquine is a potent myocardial depressant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0598