Learn More
Medchemexpress LLC Didodecylamine | 3007-31-6 | MFCD00041918 | >98.0% | 353.67 g/mol | C24H51N | 100 G
SDP
Supplier:  Medchemexpress LLC HYW142525100G
Encompass_Preferred
Didodecylamine (C24H51N, CAS 3007-31-6) is a long-chain secondary amine used as a reagent and amphiphilic intermediate in organic synthesis, surfactant research, and formulation development. It is supplied in research-grade purity for analytical and preparative applications and should be handled and stored according to its SDS.
Catalog No. 50-002-83044