Learn More
Selleck Chemical LLC Didox E2939-25MG
SDP
Supplier:  Selleck Chemical LLC E293925MG
Encompass_Preferred
Didox (3 4-dihydroxy-benzohydroxamic acid) is a synthetic ribonucleotide reductase (RR) inhibitor derived from polyhydroxy-substituted benzohydroxamic acid Didox inhibits proliferation of resistant breast cancer cells
Catalog No. 50-000-40403