Learn More
Medchemexpress LLC Dids sodium salt | 67483-13-0 | 99.1% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYD00865MG
Encompass_Preferred
DIDS sodium salt (MDL101114ZA) is a dual inhibitor of ABCA1 and VDAC1. It also inhibits RAD51, specifically RAD51-mediated homologous pairing and strand exchange reactions. DIDS inhibits anion exchange and binding to red blood cell membranes, inhibits the activation of caspase-3 and -9, and can be used in cancer research.
  • Dual inhibitor of ABCA1 and VDAC1
  • Inhibits RAD51-mediated homologous pairing and strand exchange reactions
  • Inhibits anion exchange and binding to red blood cell membranes
  • Inhibits the activation of caspase-3 and -9
  • Can be used in cancer research
Catalog No. 50-004-10653