Learn More
Grainger Die Grinder 1/4 In 3.5 A
Supplier:  Grainger 14F111
Encompass_Preferred
The Makita 3.5 Amp Die Grinder is a powerful, compact tool for grinding, sanding, honing, deburring, and general purpose grinding. It has a slim body design at only 5-1/4 inches long for working in tight spaces. The motor delivers 21,000 RPM for quick material removal. It has a variable speed control dial that allows the user to match the speed to the application. The grinder accepts a wide range of accessories including mounted stones, rotary files, abrasive cutoff wheels, and abrasive discs. It comes with a wheel guard, side handle, wrench, and case for safety and convenience. This die grinder is durable, reliable, and made for professional use.
Catalog No. 50-275-3604