Learn More
Medchemexpress LLC Diethyl (4-bromobutyl)phosphonate | 63075-66-1 | MFCD05863820 | >90.0% | C8H18BrO3P | 500 MG

Supplier: Medchemexpress LLC HY140331500MG
Diethyl (4-bromobutyl)phosphonate is a diethyl ω-bromoalkylphosphonate, a chemical compound with the formula C8H18BrO3P. It has a CAS number of 63075-66-1 and is identified by the MFCD number MFCD05863820. It typically appears as a colorless to light yellow clear liquid. It is known to cause skin and serious eye irritation.
- Can be used to synthesize new quinoquinonephosphonic acid esters.
- Molecular weight is 273.10 g/mol.
- Purity is typically greater than 90.0%. (Note: Purity value obtained from a different supplier's datasheet, not MedChemExpress.)
- Storage recommended in a cool and dark place, under an inert atmosphere, or at room temperature.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.