Learn More
Medchemexpress LLC Diethyl 4-bromobutylphosphonate | 63075-66-1 | C8H18BrO3P | 1 G
SDP
Supplier:  Medchemexpress LLC HY1403311G
Encompass_Preferred
Diethyl 4-bromobutylphosphonate is a diethyl ω-bromoalkylphosphonate that can be used to synthesize new quinoquinonephosphonic acid esters. This product is suitable for various laboratory applications.
  • Used to synthesize new quinoquinonephosphonic acid esters
Catalog No. 50-003-56800