Learn More
Medchemexpress LLC Diethyl bipy55 DC 10g | 1762-46-5 | 300.31 g·mol-1 | C16H16N2O4 | 10 G
SDP
Supplier:  Medchemexpress LLC HY11689310G
Encompass_Preferred
Diethyl bipy55'DC (Diethyl [2,2'-bipyridine]-5,5'-dicarboxylate) is a small-molecule inhibitor of collagen prolyl 4-hydroxylases (CP4Hs) used in research to modulate collagen hydroxylation and study antifibrotic and anti-metastatic mechanisms. Supplied as a research-grade powder for laboratory use, it is intended for in vitro and analytical applications only.
  • Inhibits collagen prolyl 4-hydroxylase (CP4H).
  • Used to study collagen modification, fibrosis, and metastasis.
  • Molecular formula C16H16N2O4; molecular weight 300.31 g·mol-1.
  • Identified by CAS 1762-46-5.
  • Supplied as a powder for research use only.
Catalog No. 50-002-27664