Learn More
Medchemexpress LLC Diethyl bipy55 DC 5g | 1762-46-5 | 300.31 g/mol | C16H16N2O4 | 5 G

Supplier: Medchemexpress LLC HY1168935G
Diethyl [2,2'-bipyridine]-5,5'-dicarboxylate is an organic research reagent used as a ligand precursor and in studies involving bipyridine derivatives. It is supplied as a light yellow to yellow solid for research use only and is not for human or veterinary use.
- High purity: ≥98%.
- Light yellow to yellow solid appearance.
- Molecular formula: C16H16N2O4 (source-dependent).
- Molecular weight: 300.31 g/mol (source-dependent).
- Storage: powder at -20°C; in solvent at -80°C.
- Solubility: DMSO 50 mg/mL; ethanol 5 mg/mL (may require ultrasonic).
- Available in multiple pack sizes, including 5 g.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.