Learn More
Medchemexpress LLC HY-Y0836 100mg Medchemexpress, Diethyl succinate CAS:123-25-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY0836100MG
Encompass_Preferred
Medchemexpress, HY-Y0836 100mg Diethyl succinate CAS:123-25-1 Diethyl succinate (Diethyl Butanedioate) is used at physiological pH and crosses biological membranes, incorporates into cells in tissue culture and is metabolized by the TCA cycle. Diethyl succinate is known to be non-toxic and used in fragrances and flavoring. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3551