Learn More
Medchemexpress LLC Diethyl terephthalat 10g
SDP
Supplier:  Medchemexpress LLC HYW11183010G
Encompass_Preferred
Diethyl terephthalate (Terephthalic acid) is a monomer in the synthesis of polyethylene terephthalate (PET) material Diethyl terephthalate is usually used as a substrate for isolating PET-degrading strains[1]
Catalog No. 50-002-06646