Learn More
Medchemexpress LLC Diethylamine nonoate diethylammonium salt | 372965-00-9 | MFCD00216152 | 98.0% | 206.29 g/mol | C8H22N4O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1319255MG
Encompass_Preferred
Diethylamine nonoate diethylammonium salt is a nitric oxide donor supplied as a white to light-brown solid for research use. It is used in biological studies as an NO donor with reported antimicrobial activity and applications in tissue preservation research.
  • Nitric oxide donor for biological research.
  • Demonstrated antimicrobial activity against bacteria.
  • Used in tissue preservation and related studies.
  • High purity, approximately 98.0%.
  • Molecular weight 206.29 g/mol; formula C8H22N4O2.
Catalog No. 50-002-62324