Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diethylamino hydroxybenzoyl hexyl benzoate | 302776-68-7 | 99.9% | 397.51 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY1096565G

Encompass_Preferred

Diethylamino hydroxybenzoyl hexyl benzoate (DHHB) is a photostable UV-A absorber. It is intended for research use only. It absorbs UV-A radiation with a peak at 354 nm, with a molar absorption coefficient in EtOH at 25°C of 39,000 mol/dm³/cm at 354 nm. Its emission spectrum in EtOH is dependent on the excitation wavelength.

  • Photostable UV-A absorber
  • Absorbs UV-A radiation at 354 nm
  • Molar absorption coefficient: 39,000 mol/dm³/cm at 354 nm in EtOH at 25°C
  • Emission spectrum in EtOH is excitation wavelength dependent

Catalog No. 50-003-44602


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.