Learn More
Medchemexpress LLC Diethylcarbamazine | 90-89-1 | 99.8% | 199.29 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY12642A100MG
Encompass_Preferred
Diethylcarbamazine is an orally active microfilaricidal agent primarily used in the treatment of onchocerciasis and lymphatic filariasis. It reduces eosinophil trafficking to lung tissue, exhibiting anti-allergic properties. It also lowers serum levels of leptin, TNF-α, IL-6, MCP-1, glucose, insulin, and triglycerides, improving insulin resistance. This product is suitable for research concerning bronchial asthma, insulin resistance, and infections.
  • Orally active microfilaricidal agent.
  • Reduces eosinophil trafficking and anti-allergic.
  • Lowers serum leptin, TNF-α, IL-6, MCP-1, glucose, insulin, triglycerides.
  • Improves insulin resistance.
  • Enhances reactive oxygen intermediate expression by polymorphonuclear neutrophils.
  • Increases lymphocyte proliferation.
  • Inhibits actinomycetoma lesion development.
  • Useful for research on bronchial asthma, insulin resistance, and infection.
Catalog No. 50-004-36245