Learn More
Medchemexpress LLC Diethylene glycol monohexyl ether | 112-59-4 | MFCD00010703 | 190.28 g·mol^-1 | C10H22O3 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW133950500G
Encompass_Preferred
Diethylene glycol monohexyl ether is an organic glycol ether used as a solvent and biochemical assay reagent. It is the monohexyl ether derivative of diethylene glycol, offering moderate polarity and a relatively high boiling point for use in formulations, synthesis, and analytical applications.
  • Moderate polarity solvent suitable for biochemical assays.
  • Relatively high boiling point for thermal stability in reactions.
  • Good solvency for a range of polar and nonpolar compounds.
  • Liquid form that is easy to dispense and incorporate into formulations.
  • Supplied in laboratory pack sizes for research use.
Catalog No. 50-002-82840