Learn More
Medchemexpress LLC Diethylene glycol monohexyl ether | 112-59-4 | MFCD00010703 | 190.28 g·mol⁻¹ | C10H22O3 | 1 KG
SDP
Supplier:  Medchemexpress LLC HYW1339501KG
Encompass_Preferred
Diethylene glycol monohexyl ether is a colorless liquid ether (C10H22O3, MW 190.28 g·mol⁻¹) used primarily as a nonionic solvent and biochemical assay reagent. It is employed in laboratory formulations, synthesis workflows, and applications requiring a low-volatility, organic solvent with emulsifying properties.
  • Nonionic solvent suitable for formulations and coatings.
  • Useful as a reagent or solvent in biochemical assays and synthesis.
  • Liquid at room temperature with good solubility in organic solvents.
  • Provided in multiple pack sizes to support laboratory scale needs.
  • Chemical formula C10H22O3 and molecular weight 190.28 g·mol⁻¹.
Catalog No. 50-002-82849