Learn More
Medchemexpress LLC Diethylstilbestrol-d8 | 91318-10-4 | MFCD01074086 | 99.7% | 276.40 g·mol⁻¹ | C18H12D8O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY14598S5MG
Encompass_Preferred
Diethylstilbestrol-d8 is the deuterium-labeled analogue of diethylstilbestrol supplied as a solid stable-isotope internal standard for analytical and research use. It is intended as a tracer or internal standard for quantitative NMR, GC-MS, and LC-MS assays and has high isotopic purity suitable for precise analytical quantitation.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS assays.
  • High isotopic purity (≈99.7%).
  • Soluble in DMSO (≈50 mg/mL); ultrasonic warming recommended.
  • Stable solid with recommended cold storage for long-term preservation.
  • Supplied in small analytical quantities for tracer and standard applications.
Catalog No. 50-002-22292