Learn More
Medchemexpress LLC Diethyltoluamide-d7 | 1219799-37-7 | 98.5% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0978S15MG
Encompass_Preferred
Diethyltoluamide-d7 is the deuterium-labeled form of Diethyltoluamide, which is widely recognized as the primary active ingredient in insect repellents. It is formulated to offer protection against a broad spectrum of biting insects, including mosquitoes, ticks, fleas, chiggers, and leeches. This compound is intended for research purposes only.
  • Deuterium labeled diethyltoluamide.
  • Can be used as a tracer.
  • Can serve as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, such as deuterium, are incorporated into drug molecules for quantitation during drug development.
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-08046