Learn More
Strem, An Ascensus Company CAS# 557-20-0. 50g. Diethylzinc, min. 95% (10 wt% in hexanes) (Sure/Seal™ Bottle). MFCD00009021
SDP
Supplier:  Strem, An Ascensus Company 30302950G
Encompass
CAS# 557-20-0. 50g. Diethylzinc, min. 95% (10 wt% in hexanes) (Sure/Seal™ Bottle). MFCD00009021. Molecular Weight: 123.49. Molecular Formula: Zn(C2H5)2. Color/form: colorless liq. Strem# 30-3029. http://www.strem.com/catalog/v/30-3029/
Catalog No. 50-901-11384