Learn More
Strem, An Ascensus Company Diethylzinc, min. 95% 100g
SDP
Supplier:  Strem, An Ascensus Company 933030100G
Encompass
CAS# 557-20-0. 100g. Diethylzinc, min. 95%. MFCD00009021. Molecular Weight: 123.49. Molecular Formula: Zn(C2H5)2. Color/form: colorless liq. Strem# 93-3030. http://www.strem.com/catalog/v/93-3030/
Catalog No. 50-901-14982