Learn More
Medchemexpress LLC HY-109085 5mg Medchemexpress, Difamilast CAS:937782-05-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1090855MG
Encompass_Preferred
Medchemexpress, HY-109085 5mg Difamilast CAS:937782-05-3 Difamilast (OPA-15406) is a topical, selective and nonsteroidal phosphodiesterase-4 (PDE4) inhibitor with particularly efficient inhibition of subtype B (IC50=11.2 nM). Difamilast can be used for the research of mild to moderate atopic dermatitis (AD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0763