Learn More
Medchemexpress LLC Difamilast | 937782-05-3 | 99.4% | 446.44 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10908510MG
Encompass_Preferred
Difamilast (OPA-15406) is a topical, selective, and nonsteroidal phosphodiesterase-4 (PDE4) inhibitor, demonstrating particularly efficient inhibition of subtype B with an IC50 of 11.2 nM. It is utilized in the research of mild to moderate atopic dermatitis (AD).
  • Topical, selective, and nonsteroidal phosphodiesterase-4 (PDE4) inhibitor
  • Efficiently inhibits PDE4 subtype B (IC50=11.2 nM)
  • Used in research for mild to moderate atopic dermatitis (AD)
  • Exerts inhibitory effects on PDE2
  • Improves skin condition in relevant animal models of AD
Catalog No. 50-003-43263