Learn More
Medchemexpress LLC Difamilast | 937782-05-3 | 99.4% | 446.44 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10908550MG
Encompass_Preferred
Difamilast (OPA-15406) is a topical, selective, and nonsteroidal phosphodiesterase-4 (PDE4) inhibitor with efficient inhibition of subtype B (IC50=11.2 nM). It can be used for the research of mild to moderate atopic dermatitis (AD).
  • Highly selective for inhibition of PDE4 subtype B
  • Improves skin condition in relevant animal models of AD
Catalog No. 50-003-45095