Learn More
Chiral Technologies, Inc. DIFFINITY RAPIDTIP 96/PK UNIV
Supplier:  Chiral Technologies, Inc. RT025096
Encompass
Diffinity RapidTip (96) for PCR Purification - 1x96 rack of tips (Compatible with universal pipets) - Purify 25ul PCR sample in ONE minute - Removes > 90% dNTPs, primers and primer-dimers - Retain 80-90% of PCR amplicon
Catalog No. 50-103-8713