Learn More
Perkin Elmer US LLC Diffuse Reflection Multi-Purpose Sample Holder
Supplier:  Perkin Elmer US LLC L1201865
Encompass_Preferred
The PerkinElmer Diffuse Reflectance Accessory provides a convenient and sensitive method for analyzing opaque or highly scattering solid samples such as pharmaceuticals, food products, soappowders, coal, clay, soil, paper, painted surfaces, polymer foam and catalysts. This accessory is compatible with the Spectrum One/100/400/65, Spectrum Two, and Spectrum 3/Frontier spectometers.
Catalog No. 50-211-3393