Learn More
Medchemexpress LLC HY-128171 5mg Medchemexpress, Diflapolin CAS:724453-98-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1281715MG
Encompass_Preferred
Medchemexpress, HY-128171 5mg Diflapolin CAS:724453-98-9 Diflapolin is a highly active dual 5-lipoxygenase-activating protein (FLAP)/soluble epoxide hydrolase (sEH) inhibitor with marked anti-inflammatory efficacy and high target selectivity. Diflapolin inhibits 5-LOX product formation in intact human monocytes and neutrophils with IC50s of  30 and 170 nM, respectively, and suppressed the activity of isolated sEH (IC50=20 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1155