Learn More
Medchemexpress LLC Diflorasone diacetate | 33564-31-7 | MFCD00079159 | 99.9% | 494.52 g·mol⁻¹ | C26H32F2O7 | 10 MG

Supplier: Medchemexpress LLC HY10796110MG
Diflorasone diacetate is a synthetic corticosteroid research compound used to study anti-inflammatory effects and corticosteroid-responsive dermatoses. It is provided for laboratory use as a solid or as DMSO solutions suitable for in vitro assays.
- High purity (99.9%)
- White to off-white solid appearance
- Available as small solid packs and 10 mM solution in DMSO
- Chemical formula C26H32F2O7; molecular weight 494.52 g·mol⁻¹
- Stored sealed at 4°C; in solvent store at -80°C for long-term
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.