Learn More
Selleck Chemical LLC Diflorasone diacetate E4941-25MG
SDP
Supplier:  Selleck Chemical LLC E494125MG
Encompass_Preferred
Diflorasone diacetate(Florone) is a steroid anti-inflammatory drug an inhibitor of TARC (thymus and activation regulated chemokine CCL17) in the dinitrochlorobenzene (DNCB) model It can be used in the treatment of allergic and inflammatory skin diseases such as atopic dermatitis and allergic contact dermatitis
Catalog No. 50-000-43258