Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Difloxacin-d3 hydrochloride | 1173021-89-0 | 99.0% | 10 MG
SDP

Supplier:  Medchemexpress LLC HY121272AS110MG

Encompass_Preferred

Difloxacin-d3 hydrochloride is a deuterium-labeled hydrochloride salt of difloxacin, provided as an isotopically labeled reference standard for analytical and research applications. It is supplied as a high-purity white to off-white solid suitable for use as an internal standard in mass spectrometry and quantitative assays.

  • Deuterium-labeled internal standard for analytical assays.
  • Hydrochloride salt, white to off-white solid.
  • Purity 99.0%.
  • Molecular weight 438.87 g/mol.
  • Chemical formula C21H17D3ClF2N3O3.
  • Available in small milligram quantities for research use.
  • Store sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-31382


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.