Learn More
Medchemexpress LLC Diflubenzuron | 35367-38-5 | 98.7% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1973100MG
Encompass_Preferred
Diflubenzuron is an insecticide that exhibits both larvicidal and ovicidal activities. Its mechanism of action involves inhibiting the synthesis of chitin in insects, which interferes with their molting process and ultimately leads to their death. It is for research use only.
  • Insecticide with larvicidal and ovicidal activities
  • Inhibits chitin synthesis
  • Affects insect molting
Catalog No. 50-004-09674