Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diflufenican | 83164-33-4 | 99.8% | 394.29 | C19H11F5N2O2 | 100 MG
SDP

Supplier:  Medchemexpress LLC HYW040206100MG

Encompass_Preferred

Diflufenican is a contact, selective herbicide used in research to control certain broad-leaved weeds. It is supplied as an off-white to light yellow solid with high reported purity and shows good solubility in DMSO; storage recommendations differ for powder and solutions.

  • Product form: solid (off-white to light yellow)
  • Purity: 99.77%
  • Cas number: 83164-33-4
  • Chemical formula and molecular weight: C19H11F5N2O2; 394.29
  • Solubility: DMSO 66.67 mg/mL; formulation example 10% DMSO in corn oil ≥2.5 mg/mL
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
  • Packaging sizes: 100 mg, 250 mg, 500 mg, 1 g

Catalog No. 50-002-09326


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.