Learn More
Medchemexpress LLC Diflufenican | 83164-33-4 | MFCD01311804 | 99.8% | 394.29 | C19H11F5N2O2 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW040206
Diflufenican is a contact, selective herbicide supplied as a solid analytical standard intended for research and analytical use. The product is offered as a 250 mg solid with high reported purity and defined storage recommendations.
- High purity analytical standard (99.77%).
- CAS number 83164-33-4.
- Molecular weight 394.29 g/mol.
- Molecular formula C19H11F5N2O2.
- Supplied as a solid with recommended storage at -20°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.