Learn More
Cayman Chemical DIfluprednate 10mg
SDP
Supplier:  Cayman Chemical 2377610 MG
Encompass_Preferred
A synthetic glucocorticoid (Ki = 78 pM for the glucocorticoid receptor); increases intraocular pressure in sheep following ocular administration of a 0.05% w/v solution
Catalog No. 50-299-5256