Learn More
Medchemexpress LLC Difluprednate | 23674-86-4 | 99.6% | 508.55 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1756950MG
Encompass_Preferred
Difluprednate is a glucocorticoid receptor (GC receptor) agonist with a Ki of 0.78 nM. Its active metabolite, 21-deacetylated difluprednate (DFB), is a competitive agonist of GC receptors with a Ki of 0.061 nM. It binds to GC receptors through metabolism to DFB, regulating the transcription of inflammatory mediator genes, thereby exerting anti-inflammatory activity. It can be used for research related to ocular inflammation, such as postoperative inflammation and anterior uveitis.
  • Glucocorticoid receptor (GC receptor) agonist
  • Active metabolite DFB is a competitive agonist of GC receptors
  • Regulates transcription of inflammatory mediator genes
  • Exerts anti-inflammatory activity
  • Used for research related to ocular inflammation, such as postoperative inflammation and anterior uveitis
Catalog No. 50-003-87500