Learn More
Sigma Aldrich Fine Chemicals Biosciences DIG Easy Hyb, 500 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11603558001
Encompass
DIG Easy Hyb can be used in all membrane hybridization applications using nonradioactive (e.g., digoxigenin-labeled) nucleic acid probes. DIG Easy Hyb is ready-made, and can be used without any additions. It is specifically designed for use with DIG kits.
Catalog No. NC1511595