Learn More
Sheldon Manufacturing Inc DIG LAB INCUBATOR SGL CHAMBER
SDP
Supplier:  Sheldon Manufacturing Inc SLM1122/DUP
Encompass_Preferred
LABORATORY INCUBATOR, SIDE BY SIDE, 11 CU FT, SS INTERIOR, INNER GLASS DOOR, OUTLET, ACCESS PORT, 115V. SHEL LAB general purpose incubators deliver a degree of temperature uniformity usually found only in more expensive, application specific models. This high level of performance is due to a precise microprocessor controller and a unique warm air jacket design. Triple wall construction allows heated air to circulate within an open plenum between the inner chamber and exterior walls. This design does not require blowers or fans within the incubator chamber where samples are susceptible to cross contamination and the drying effects of convection. Five strategically placed heating elements, including a heated outer door, ensure uniform heating, even when the incubator chamber is fully loaded. More information available at www.shellab.com
Catalog No. NC0962983