Learn More
Cole-Parmer Traceable Extra-Large Digit Digital Timer with Calibration
Supplier:  Cole-Parmer 9446107
Encompass_Preferred
Simple operation and memory function make this the ideal timer for repetitive timing applications. Intuitive three-button operation lets you quickly program and start your timing application. Automatic Bounce back™ memory function recalls your set time for repetitive timing operations. Also counts up in stopwatch mode. Built-in easel stand, pocket clip, and magnetic back give you multiple mounting options. Traceable Products have an individually-numbered Traceable Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Catalog No. NC1470665