Learn More
Cole-Parmer Fowler 54-101-150-2 Xtra-Value CAL Electronic Caliper, 6"/150 mm
Supplier:  Cole-Parmer 9723134
Encompass_Preferred
Caliper features a stainless steel frame, large easy-to-read display, 4-way measuring, resolution of 0.0005"/0.01 mm, absolute/incremental measurement and accuracy of 0.0001"/0.02".
Catalog No. NC9266915