Learn More
Research Products International Corp Digital Gel Imaging and Documentation System
SDP
Supplier:  Research Products International Corp 970105
Encompass
This economical system provides true low cost image acquisition and documentation of gels, plates and membranes. The system combines a 5.1 mega pixel high-resolution digital color camera with a compact enclosure and UV filter. Custom designed software links the camera to an existing computer via a USB interface. Software compatible with Window 98, ME and 2000.
System includes digital color camera, software, hood enclosure, UV filter, diopter and universal power supply.
Transiluminator sold separately.
Catalog No. 50-197-6814
Add to Cart
Request a Quote