Learn More
BRANSON ULTRASONICS CORPORATION (3P) DIGITAL HEATED ULTRASONIC BATH
Supplier:  BRANSON ULTRASONICS CORPORATION (3P) CPX952118R
Encompass_Preferred
Branson Ultrasonics CPXH Series Ultrasonic Cleaning Bath, Model: CPX1800H, Capacity: 1.9L (0.5 gal.), Dimensions Interior: 15 x 14 x 10cm (6 x 5.5 x 4 in.), Voltage: 120V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover, Plastic Housing, Stainless Steel Tank, Timer: 90 min.
Catalog No. NC3721471