Learn More
Cole-Parmer Stuart SMP10 Digital Melting Point Apparatus, 240 VAC
Supplier:  Cole-Parmer 0301141
Encompass_Preferred
Designed for maximum safety and ease of use. Select, measure, and display the temperature digitally, ensuring accuracy and avoiding the need for a glass thermometer. Test two samples simultaneously, viewing the melt via a magnifying lens with built-in illumination. Extend the back feet for the optimal viewing angle.
Set a plateau temperature for initial warm-up — unattended warm-up time frees up the user until the plateau temperature is reached. Heating rate continues at 2°C after the warm-up, slowly reaching the melt temperature for greater accuracy.
Catalog No. 50-196-0404