Learn More
RWD LIFE SCIENCE INC DIGITAL OPERATING MICROSCOPE
SDP
Supplier:  RWD LIFE SCIENCE INC DOM1001
Encompass_Preferred
DOM-1001 is a 16 Million Pixels high-end integrated digital microscope, and enables real-time record, collect, and share high-definition 4K images and videos(1080P). Multiple people can view samples simultaneously, supporting the whole process of taking pictures and videos. it is widely applied in Life science: dental, animal surgery, tumor, morphological observation, embryologic histology, education, Industry and Manufacturing: electronics and semiconductor, metals, medical, new energy, etc.
Catalog No. NC3404316