Learn More
Cole-Parmer Digital Pressure Gauge w/ Backlight, 0-500 psi
Supplier:  Cole-Parmer 6833421
Encompass_Preferred
Ashcroft DG25 Digital Pressure Gauge with Backlight, 0 to 500 psi, Stainless steel sensor and nine engineering units make this gauge suitable for many applications±0.5% full-scale accuracyIP67 rated caseEasy-to-read, 5-digit display with 20-segment bar graph0 to 500 psi range
Catalog No. NC0707435
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov