Learn More
Andwin Scientific DIGITAL THERMOMETER MODEL51
SDP
Supplier:  Andwin Scientific 61221038
Encompass_Preferred
The fluke digital thermometer is made to be utilized with Type-K, Type-J, Type-T, or Type-E thermocouples. It is able to read temperature in units of Celsius, Farenheit, and Kelvin. This product features a relative time clock that gives a time reference for minimum, maximum, and average reading. The thermometer also has a hold button that conveniently permits the freezing of the reading for viewing purposes. It has a dimension of 17.3 x 8.6 x 3.8 cm (613/16 x 33/8 x 1 1/2").
Catalog No. NC9849802